Learn More
Biomatik Corporation A Disintegrin And Metalloproteinase With Thrombospondin 5 (Adamts5) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU21598
Encompass_Preferred
A Disintegrin And Metalloproteinase With Thrombospondin 5 (Adamts5) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to A Disintegrin And Metalloproteinase With Thrombospondin 5 (ADAMTS5). The A Disintegrin And Metalloproteinase With Thrombospondin 5 (Adamts5) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Arg267~Asn622. Applications: WB, IHC, ICC, IP. Target Synonyms: A Disintegrin And Metalloproteinase With Thrombospondin 5 (ADAMTS5). Accession Number: Q6TY19. Gene ID: 304135. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9212