Learn More
Biomatik Corporation Cluster Of Differentiation 55 (Cd55) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27855
Encompass_Preferred
Cluster Of Differentiation 55 (Cd55) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Cluster Of Differentiation 55 (CD55). The Cluster Of Differentiation 55 (Cd55) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Pro47~Glu286. Applications: WB, IHC, ICC, IP. Target Synonyms: Cluster Of Differentiation 55 (CD55). Accession Number: Q61475. Gene ID: 13136. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9526