Learn More
Biomatik Corporation Cathepsin K (Ctsk) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU28229
Encompass_Preferred
Cathepsin K (Ctsk) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Cathepsin K (CTSK). The Cathepsin K (Ctsk) Polyclonal Antibody has been validated for the following applications: WB, ICC, IHC-P, IHC-F, ELISA. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Leu54~Ala317. Applications: WB, ICC, IHC-P, IHC-F, ELISA. Target Synonyms: Cathepsin K (CTSK). Accession Number: P43235. Gene ID: 1513. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9540