Learn More
Biomatik Corporation Adiponectin Receptor 2 (Adipor2) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU28507
Encompass_Preferred
Adiponectin Receptor 2 (Adipor2) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Adiponectin Receptor 2 (ADIPOR2). The Adiponectin Receptor 2 (Adipor2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Met1~Gly147. Applications: WB, IHC, ICC, IP. Target Synonyms: Adiponectin Receptor 2 (ADIPOR2). Accession Number: Q3YAF7. Gene ID: 312670. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9548