Learn More
Biomatik Corporation Collagen Type Vi Alpha 3 (Col6A3) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU24611
Encompass_Preferred
Collagen Type Vi Alpha 3 (Col6A3) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Collagen Type VI Alpha 3 (COL6a3). The Collagen Type Vi Alpha 3 (Col6A3) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Ala463~Leu624. Applications: WB, IHC, ICC, IP. Target Synonyms: Collagen Type VI Alpha 3 (COL6a3). Accession Number: P12111. Gene ID: 1293. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9354