Learn More
Biomatik Corporation Coagulation Factor Xi (F11) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU25212
Encompass_Preferred
Coagulation Factor Xi (F11) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Coagulation Factor XI (F11). The Coagulation Factor Xi (F11) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Ile275~Gly516. Applications: WB, IHC, ICC, IP. Target Synonyms: Coagulation Factor XI (F11). Accession Number: Q6TUF8. Gene ID: 290757. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9383