Learn More
Biomatik Corporation Signal Transducer And Activator Of Transcription 1 (Stat1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU25401
Encompass_Preferred
Signal Transducer And Activator Of Transcription 1 (Stat1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Signal Transducer And Activator Of Transcription 1 (STAT1). The Signal Transducer And Activator Of Transcription 1 (Stat1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Val343~Arg602. Applications: WB, IHC, ICC, IP. Target Synonyms: Signal Transducer And Activator Of Transcription 1 (STAT1). Accession Number: P42224. Gene ID: 6772. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9393