Learn More
Biomatik Corporation Cluster Of Differentiation 42D (Cd42D) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26188
Encompass_Preferred
Cluster Of Differentiation 42D (Cd42D) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Cluster of Differentiation 42d (CD42d). The Cluster Of Differentiation 42D (Cd42D) Polyclonal Antibody has been validated for the following applications: WB, FCM. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Gln17~Pro242. Applications: WB, FCM. Target Synonyms: Cluster of Differentiation 42d (CD42d). Accession Number: P40197. Gene ID: 2814. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9445