Learn More
Biomatik Corporation Small Nuclear Ribonucleoprotein Polypeptide D1 (Snrpd1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26473
Encompass_Preferred
Small Nuclear Ribonucleoprotein Polypeptide D1 (Snrpd1) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Small Nuclear Ribonucleoprotein Polypeptide D1 (SNRPD1). The Small Nuclear Ribonucleoprotein Polypeptide D1 (Snrpd1) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Arg119. Applications: WB, IHC. Target Synonyms: Small Nuclear Ribonucleoprotein Polypeptide D1 (SNRPD1). Accession Number: P62314. Gene ID: 6632. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9460