Learn More
Biomatik Corporation Elastin Microfibril Interface Located Protein 1 (Emilin1) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27935
Encompass_Preferred
Elastin Microfibril Interface Located Protein 1 (Emilin1) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Elastin Microfibril Interface Located Protein 1 (EMILIN1). The Elastin Microfibril Interface Located Protein 1 (Emilin1) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Leu684~Gln899. Applications: WB, IHC, ICC, IP. Target Synonyms: Elastin Microfibril Interface Located Protein 1 (EMILIN1). Accession Number: D3Z9E1. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9532