Learn More
Biomatik Corporation Coagulation Factor Xi (F11) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU29118
Encompass_Preferred
Coagulation Factor Xi (F11) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Coagulation Factor XI (F11). The Coagulation Factor Xi (F11) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Pro157~Arg389. Applications: WB, IHC, ICC, IP. Target Synonyms: Coagulation Factor XI (F11). Accession Number: Q91Y47. Gene ID: 109821. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9568