Learn More
Biomatik Corporation Interleukin 1 Beta (Il1B) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29272
Encompass_Preferred
Interleukin 1 Beta (Il1B) Monoclonal Antibody is a Rat (Rattus norvegicus) Monoclonal Antibody to Interleukin 1 Beta (IL1b). The Interleukin 1 Beta (Il1B) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Rat (Rattus norvegicus). Immunogen: Val117~Ser268. Applications: WB, IHC, ICC, IP. Target Synonyms: Interleukin 1 Beta (IL1b). Accession Number: Q63264. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9578