Learn More
Biomatik Corporation Collagen Type I Alpha 1 (Col1A1) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29527
Encompass_Preferred
Collagen Type I Alpha 1 (Col1A1) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Collagen Type I Alpha 1 (COL1a1). The Collagen Type I Alpha 1 (Col1A1) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Arg253~Ala471. Applications: WB, IHC, ICC, IP. Target Synonyms: Collagen Type I Alpha 1 (COL1a1). Accession Number: P02452. Gene ID: 1277. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9599