Learn More
Biomatik Corporation Arginase (Arg) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29831
Encompass_Preferred
Arginase (Arg) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Arginase (ARG). The Arginase (Arg) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Met1~Lys322. Applications: WB, IHC, ICC, IP. Target Synonyms: Arginase (ARG). Accession Number: P05089. Gene ID: 383. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9615