Learn More
Biomatik Corporation Neuregulin 4 (Nrg4) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29862
Encompass_Preferred
Neuregulin 4 (Nrg4) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Neuregulin 4 (NRG4). The Neuregulin 4 (Nrg4) Monoclonal Antibody has been validated for the following applications: WB. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Met1~Phe62. Applications: WB. Target Synonyms: Neuregulin 4 (NRG4). Accession Number: Q8WWG1. Gene ID: 145957. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9618