Learn More
Biomatik Corporation Klotho Beta (Klb) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU21888
Encompass_Preferred
Klotho Beta (Klb) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Klotho Beta (KLb). The Klotho Beta (Klb) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Val517~Ser636. Applications: WB, IHC. Target Synonyms: Klotho Beta (KLb). Accession Number: Q86Z14. Gene ID: 152831. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9228