Learn More
Biomatik Corporation Relaxin 2 (Rln2) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU23018
Encompass_Preferred
Relaxin 2 (Rln2) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Relaxin 2 (RLN2). The Relaxin 2 (Rln2) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Met1~Cys185. Applications: WB, IHC, ICC, IP. Target Synonyms: Relaxin 2 (RLN2). Accession Number: P04090. Gene ID: 6019. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9282