Learn More
Biomatik Corporation Peroxisome Proliferator Activated Receptor Alpha (Ppara) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU26812
Encompass_Preferred
Peroxisome Proliferator Activated Receptor Alpha (Ppara) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Peroxisome Proliferator Activated Receptor Alpha (PPARa). The Peroxisome Proliferator Activated Receptor Alpha (Ppara) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: Pro61~Ala170. Applications: WB, IHC, ICC, IP. Target Synonyms: Peroxisome Proliferator Activated Receptor Alpha (PPARa). Accession Number: Q07869. Gene ID: 5465. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9472