Learn More
Biomatik Corporation Mucin 5 Subtype B (Muc5B) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27377
Encompass_Preferred
Mucin 5 Subtype B (Muc5B) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Mucin 5 Subtype B (MUC5B). The Mucin 5 Subtype B (Muc5B) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Ala75~Leu295. Applications: WB, IHC, ICC, IP. Target Synonyms: Mucin 5 Subtype B (MUC5B). Accession Number: Q8K1G6. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9507