Learn More
Biomatik Corporation Interleukin 1 Beta (Il1B) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU27640
Encompass_Preferred
Interleukin 1 Beta (Il1B) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Interleukin 1 Beta (IL1b). The Interleukin 1 Beta (Il1B) Polyclonal Antibody has been validated for the following applications: WB. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Val117~Ser268. Applications: WB. Target Synonyms: Interleukin 1 Beta (IL1b). Accession Number: Q63264. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9518