Learn More
Biomatik Corporation Connexin 43 (Cx43) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU28207
Encompass_Preferred
Connexin 43 (Cx43) Polyclonal Antibody is a Rat (Rattus norvegicus) Polyclonal Antibody to Connexin 43 (CX43). The Connexin 43 (Cx43) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Rat (Rattus norvegicus). Immunogen: Ser180~Ile382. Applications: WB, IHC, ICC, IP. Target Synonyms: Connexin 43 (CX43). Accession Number: P08050. Gene ID: 24392. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9538