Learn More
Biomatik Corporation Leptin Receptor (Lepr) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU28681
Encompass_Preferred
Leptin Receptor (Lepr) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Leptin Receptor (LEPR). The Leptin Receptor (Lepr) Polyclonal Antibody has been validated for the following applications: WB, IHC. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Leu22~Lys203. Applications: WB, IHC. Target Synonyms: Leptin Receptor (LEPR). Accession Number: P48356. Gene ID: 16847. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9550