Learn More
Biomatik Corporation Macrophage Inflammatory Protein 3 Alpha (Mip3A) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29188
Encompass_Preferred
Macrophage Inflammatory Protein 3 Alpha (Mip3A) Monoclonal Antibody is a Human (Homo sapiens) Monoclonal Antibody to Macrophage Inflammatory Protein 3 Alpha (MIP3a). The Macrophage Inflammatory Protein 3 Alpha (Mip3A) Monoclonal Antibody has been validated for the following applications: WB, IHC. Host: Mouse. Species: Human (Homo sapiens). Immunogen: Ala27~Met96. Applications: WB, IHC. Target Synonyms: Macrophage Inflammatory Protein 3 Alpha (MIP3a). Accession Number: P78556. Gene ID: 6364. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9574