Learn More
Biomatik Corporation Peptidase Inhibitor 16 (Pi16) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU21109
Encompass_Preferred
Peptidase Inhibitor 16 (Pi16) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Peptidase Inhibitor 16 (PI16). The Peptidase Inhibitor 16 (Pi16) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Leu21~Lys468. Applications: WB, IHC, ICC, IP. Target Synonyms: Peptidase Inhibitor 16 (PI16). Accession Number: Q9ET66. Gene ID: 74116. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9191