Learn More
Biomatik Corporation Recombinant Endoglin (ENG), 100UG
Supplier:  Biomatik Corporation RPU41258
Encompass_Preferred
Recombinant Human Endoglin (ENG) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Endoglin (ENG). Target Synonyms: CD105; END; HHT1; ORW; ORW1; Osler-Rendu-Weber Syndrome 1. Accession Number: P17813. Gene ID: 2022. Expression Region: Thr336~Phe450. Tag Info: N-terminal His Tag. Theoretical MW: 16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5539