Learn More
Biomatik Corporation Recombinant Endothelin 1 (EDN1), 100UG
Supplier:  Biomatik Corporation RPU41269
Encompass_Preferred
Recombinant Human Endothelin 1 (EDN1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Endothelin 1 (EDN1). Target Synonyms: ET1; PPET1; Preproendothelin-1; Big endothelin-1. Accession Number: P05305. Gene ID: 1906. Expression Region: Cys53~Ser90. Tag Info: N-terminal His Tag. Theoretical MW: 5kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5540