Learn More
Biomatik Corporation Recombinant Endothelin 1 (EDN1), 100UG
Catalog No. 501505542
Encompass_Preferred
Recombinant Rat Endothelin 1 (EDN1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Endothelin 1 (EDN1). Target Synonyms: ET1; PPET1; Preproendothelin-1; Big endothelin-1. Accession Number: P22388. Gene ID: 24323. Expression Region: Ser54~His202. Tag Info: N-terminal His Tag. Theoretical MW: 19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5542 Supplier Biomatik Corporation Supplier No. RPU41272
Recombinant Rat Endothelin 1 (EDN1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Endothelin 1 (EDN1). Target Synonyms: ET1; PPET1; Preproendothelin-1; Big endothelin-1. Accession Number: P22388. Gene ID: 24323. Expression Region: Ser54~His202. Tag Info: N-terminal His Tag. Theoretical MW: 19kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.