Learn More
Biomatik Corporation Recombinant Epidermal Growth Factor (EGF), 100UG
Supplier:  Biomatik Corporation RPU41295
Encompass_Preferred
Recombinant Pig Epidermal Growth Factor (EGF) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Pig (Sus scrofa; Porcine). Target Name: Epidermal Growth Factor (EGF). Target Synonyms: URG; Beta-Urogastrone; Pro-epidermal growth factor. Accession Number: Q00968. Gene ID: 397083. Expression Region: Ala45~His275. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5544