Learn More
Biomatik Corporation Recombinant Fibrinogen Like Protein 2 (FGL2), 100UG
Supplier:  Biomatik Corporation RPU41428
Encompass_Preferred
Recombinant Human Fibrinogen Like Protein 2 (FGL2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Fibrinogen Like Protein 2 (FGL2). Target Synonyms: T49; PT49; Fibroleukin. Accession Number: Q14314. Gene ID: 10875. Expression Region: Asn24~Pro439. Tag Info: N-terminal His Tag. Theoretical MW: 51kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5546