Learn More
Biomatik Corporation Recombinant Fibronectin (FN), 100UG
Supplier:  Biomatik Corporation RPU41461
Encompass_Preferred
Recombinant Mouse Fibronectin (FN) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Fibronectin (FN). Target Synonyms: FN1; CIG; FINC; LETS; MSF; GFND2; Anastellin; Migration-Stimulating Factor; Cold-Insoluble Globulin; Large, External, Transformation-Sensitive Protein. Accession Number: P11276. Gene ID: 14268. Expression Region: Gly2117~Leu2236. Tag Info: N-terminal His Tag. Theoretical MW: 18kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5550