Learn More
Biomatik Corporation Recombinant Ficolin 3 (FCN3), 100UG
Supplier:  Biomatik Corporation RPU41493
Encompass_Preferred
Recombinant Human Ficolin 3 (FCN3) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Ficolin 3 (FCN3). Target Synonyms: FCNH; HAKA1; Collagen/Fibrinogen Domain Containing; Hakata Antigen; Collagen/fibrinogen domain-containing lectin 3 p35. Accession Number: O75636. Gene ID: 8547. Expression Region: Glu34~Met297. Tag Info: N-terminal His Tag. Theoretical MW: 31kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5551