Learn More
Biomatik Corporation Recombinant Forkhead Box Protein C1 (FOXC1), 100UG
Supplier:  Biomatik Corporation RPU41522
Encompass_Preferred
Recombinant Mouse Forkhead Box Protein C1 (FOXC1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Forkhead Box Protein C1 (FOXC1). Target Synonyms: FOX-C1; ARA; FKHL7; FREAC3; IGDA; IHG1; IRID1; Forkhead-related protein FKHL7; Forkhead-related transcription factor 3. Accession Number: Q61572. Gene ID: 17300. Expression Region: Arg62~Lys186. Tag Info: N-terminal His Tag. Theoretical MW: 16/18kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5554