Learn More
Biomatik Corporation Recombinant Fucosidase Alpha L1, Tissue (FUCa1), 100UG
Supplier:  Biomatik Corporation RPU41530
Encompass_Preferred
Recombinant Mouse Fucosidase Alpha L1, Tissue (FUCa1) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Fucosidase Alpha L1, Tissue (FUCa1). Target Synonyms: A-FU; aFU; Alpha-L-Fucosidase; Alpha-L-fucoside fucohydrolase 1. Accession Number: Q99LJ1. Gene ID: 71665. Expression Region: Asn185~Lys447. Tag Info: N-terminal His Tag. Theoretical MW: 34kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5555