Learn More
Biomatik Corporation Recombinant Galectin 12 (GAL12), 100UG
Supplier:  Biomatik Corporation RPU41560
Encompass_Preferred
Recombinant Human Galectin 12 (GAL12) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Galectin 12 (GAL12). Target Synonyms: LGALS12; GRIP1; Lectin,Galactoside-Binding Soluble 12; Galectin-related inhibitor of proliferation. Accession Number: Q96DT0. Gene ID: 85329. Expression Region: Pro70~Leu291. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5556