Learn More
Biomatik Corporation Recombinant Galectin 3 (GAL3), 100UG
Supplier:  Biomatik Corporation RPU41565
Encompass_Preferred
Recombinant Rat Galectin 3 (GAL3) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Galectin 3 (GAL3). Target Synonyms: LGALS3; CBP35; GALBP; GALIG; MAC2; 35 kDa lectin; Carbohydrate-binding protein 35; Galactose-specific lectin 3; Galactoside-binding protein; Laminin-binding protein; Lectin L-29. Accession Number: P08699. Gene ID: 83781. Expression Region: Met1~Ile262. Tag Info: N-terminal His Tag. Theoretical MW: 28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5557