Learn More
Biomatik Corporation Recombinant Human Interferon epsilon (IFNE), 100UG
Catalog No. 501504632
Encompass_Preferred
Recombinant Human Interferon epsilon (IFNE) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: IFNE. Target Synonyms: Interferon epsilon-1. Accession Number: Q86WN2. Expression Region: 22~208aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 26.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4632 Supplier Biomatik Corporation Supplier No. RPC20162
Recombinant Human Interferon epsilon (IFNE) is a purified Recombinant Protein. Purity: >90% as determined by SDS-PAGE. Host: E. coli. Endotoxin Level: Not Tested. Species: Human (Homo sapiens). Target Name: IFNE. Target Synonyms: Interferon epsilon-1. Accession Number: Q86WN2. Expression Region: 22~208aa. Tag Info: N-Terminal 6Xhis-Tagged. Theoretical MW: 26.1kDa. Buffer: If the delivery form is liquid, the default storage buffer is Tris/PBS-based buffer, 5%-50% glycerol. If the delivery form is lyophilized powder, the buffer before lyophilization is Tris/PBS-based buffer, 6% Trehalose, pH 8.0. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.