Learn More
Biomatik Corporation Recombinant Glycocalicin (GC), 100UG
Supplier:  Biomatik Corporation RPU41693
Encompass_Preferred
Recombinant Human Glycocalicin (GC) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Glycocalicin (GC). Target Synonyms: Carbohydrate-Rich Portion of Platelet Membrane Glycoprotein Ib Alpha Polypeptide. Accession Number: P07359. Gene ID: 2811. Expression Region: Leu136~Pro276. Tag Info: N-terminal His Tag. Theoretical MW: 17/16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5561