Learn More
Biomatik Corporation Recombinant Glycoprotein 39, Cartilage (GP39), 100UG
Catalog No. 501505563
Encompass_Preferred
Recombinant Human Chitinase-3-like Protein 1 (CHI3L1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Chitinase-3-like Protein 1 (CHI3L1). Target Synonyms: GP39; CHI3-L1; YKL40; Chondrex; Chitinase 3-Like 1; Chondrocyte Protein YKL40; 39 kDa synovial protein. Accession Number: P36222. Gene ID: 1116. Expression Region: Asn112~Thr237+Ile299~Lys377. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5563 Supplier Biomatik Corporation Supplier No. RPU41707
Recombinant Human Chitinase-3-like Protein 1 (CHI3L1) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Chitinase-3-like Protein 1 (CHI3L1). Target Synonyms: GP39; CHI3-L1; YKL40; Chondrex; Chitinase 3-Like 1; Chondrocyte Protein YKL40; 39 kDa synovial protein. Accession Number: P36222. Gene ID: 1116. Expression Region: Asn112~Thr237+Ile299~Lys377. Tag Info: N-terminal His Tag. Theoretical MW: 26kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.