Learn More
Biomatik Corporation Recombinant Gremlin 1 (GREM1), 100UG
Catalog No. 501505566
Encompass_Preferred
Recombinant Mouse Gremlin 1 (GREM1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Gremlin 1 (GREM1). Target Synonyms: PIG2; CKTSF1B1; DAND2; IHG-2; DAND2; DRM; Increased in high glucose protein 2; Cell proliferation-inducing gene 2 protein; Cysteine knot superfamily 1, BMP antagonist 1. Accession Number: O70326. Gene ID: 23892. Expression Region: Lys25~Asp184. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5566 Supplier Biomatik Corporation Supplier No. RPU41746
Recombinant Mouse Gremlin 1 (GREM1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: Gremlin 1 (GREM1). Target Synonyms: PIG2; CKTSF1B1; DAND2; IHG-2; DAND2; DRM; Increased in high glucose protein 2; Cell proliferation-inducing gene 2 protein; Cysteine knot superfamily 1, BMP antagonist 1. Accession Number: O70326. Gene ID: 23892. Expression Region: Lys25~Asp184. Tag Info: N-terminal His Tag. Theoretical MW: 27kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.