Learn More
Biomatik Corporation Recombinant Growth Arrest Specific Protein 6 (GAS6), 100UG
Supplier:  Biomatik Corporation RPU41752
Encompass_Preferred
Recombinant Rat Growth Arrest Specific Protein 6 (GAS6) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: Growth Arrest Specific Protein 6 (GAS6). Target Synonyms: AXLLG; AXSF; AXL Stimulatory Factor; AXL receptor tyrosine kinase ligand. Accession Number: Q63772. Expression Region: Glu100~Gln312. Tag Info: N-terminal His Tag. Theoretical MW: 28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5568