Learn More
Biomatik Corporation Recombinant Growth Differentiation Factor 11 (GDF11), 100UG
Supplier:  Biomatik Corporation RPU41758
Encompass_Preferred
Recombinant Human Growth Differentiation Factor 11 (GDF11) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Growth Differentiation Factor 11 (GDF11). Target Synonyms: BMP11; Bone morphogenetic protein 11. Accession Number: O95390. Gene ID: 10220. Expression Region: Thr138~Asn386. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5569