Learn More
Biomatik Corporation Recombinant Growth Differentiation Factor 5 (GDF5), 100UG
Supplier:  Biomatik Corporation RPU41765
Encompass_Preferred
Recombinant Human Growth Differentiation Factor 5 (GDF5) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Growth Differentiation Factor 5 (GDF5). Target Synonyms: CDMP1; LAP4; SYNS2; BMP14; Radotermin; Cartilage-Derived Morphogenetic Protein-1; Bone morphogenetic protein 14; Lipopolysaccharide-associated protein 4. Accession Number: P43026. Gene ID: 8200. Expression Region: Ala382~Arg501. Tag Info: N-terminal His Tag. Theoretical MW: 18kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5571