Learn More
Biomatik Corporation Recombinant Heparan Sulfate Proteoglycan (HSPG), 100UG
Supplier:  Biomatik Corporation RPU41864
Encompass_Preferred
Recombinant Human Heparan Sulfate Proteoglycan (HSPG) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Heparan Sulfate Proteoglycan (HSPG). Target Synonyms: SDC2; HSPG1; SYND2; Fibroglycan; Syndecan 2; Syndecan Proteoglycan 2; Heparan Sulfate Proteoglycan 1,Cell Surface-Associated; Heparan Sulphate Proteoglycan. Accession Number: P34741. Gene ID: 6383. Expression Region: Glu19~Glu144. Tag Info: N-terminal His Tag. Theoretical MW: 23kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5576