Learn More
Biomatik Corporation Recombinant Heparan Sulfate Proteoglycan 2 (HSPG2), 100UG
Supplier:  Biomatik Corporation RPU41865
Encompass_Preferred
Recombinant Human Heparan Sulfate Proteoglycan 2 (HSPG2) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Heparan Sulfate Proteoglycan 2 (HSPG2). Target Synonyms: PLC; PRCAN; SJA; SJS; SJS1; Perlecan; Endorepellin; Schwartz-Jampel Syndrome 1; Basement membrane-specific heparan sulfate proteoglycan core protein. Accession Number: P98160. Gene ID: 3339. Expression Region: Leu4149~Ser4391. Tag Info: N-terminal His Tag. Theoretical MW: 28kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5577