Learn More
Biomatik Corporation Recombinant Hepcidin (Hepc), 100UG
Supplier:  Biomatik Corporation RPU41886
Encompass_Preferred
Recombinant Human Hepcidin (Hepc) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Hepcidin (Hepc). Target Synonyms: HAMP; HFE2B; PLTR; LEAP1; Hepcidin Antimicrobial Peptide; Liver-expressed antimicrobial peptide 1; Putative liver tumor regressor. Accession Number: P81172. Gene ID: 57817. Expression Region: Ser25~Thr84. Tag Info: N-terminal His Tag. Theoretical MW: 16kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5578