Learn More
Biomatik Corporation Recombinant Heterogeneous Nuclear Ribonucleoprotein A2/B1 (HNRPA2B1), 100UG
Supplier:  Biomatik Corporation RPU41890
Encompass_Preferred
Recombinant Human Heterogeneous Nuclear Ribonucleoprotein A2/B1 (HNRPA2B1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Heterogeneous Nuclear Ribonucleoprotein A2/B1 (HNRPA2B1). Target Synonyms: hnRNP-A2/B1; hnRNPA2B1; RA33; HnRNPA2/B1. Accession Number: P22626. Gene ID: 3181. Expression Region: Arg21~Gly103. Tag Info: N-terminal His Tag. Theoretical MW: 10/13/14kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5579