Learn More
Biomatik Corporation Recombinant High Mobility Group Protein 1 (HMG1), 100UG
Supplier:  Biomatik Corporation RPU41912
Encompass_Preferred
Recombinant Mouse High Mobility Group Protein 1 (HMGB1) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Mouse (Mus musculus). Target Name: High Mobility Group Protein 1 (HMGB1). Target Synonyms: HMG1; HMG3; SBP1; Sulfoglucuronyl Carbohydrate Binding Protein; Amphoterin; High Mobility Group Box 1 Protein. Accession Number: P63158. Gene ID: 15289. Expression Region: Met1~Glu215. Tag Info: N-terminal His Tag. Theoretical MW: 33kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5581