Learn More
Biomatik Corporation Recombinant High Molecular Weight Kininogen (HMWK), 100UG
Catalog No. 501505582
Encompass_Preferred
Recombinant Rat High Molecular Weight Kininogen (HMWK) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: High Molecular Weight Kininogen (HMWK). Target Synonyms: Williams-Fitzgerald-Flaujeac Factor; Fitzgerald Factor; HMWK-Kallikrein Factor. Accession Number: P08934. Expression Region: Leu214~Asp475. Tag Info: N-terminal His and GST Tag. Theoretical MW: 59kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5582 Supplier Biomatik Corporation Supplier No. RPU41916
Recombinant Rat High Molecular Weight Kininogen (HMWK) is a purified Prokaryotic Protein. Purity: >95% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Rat (Rattus norvegicus). Target Name: High Molecular Weight Kininogen (HMWK). Target Synonyms: Williams-Fitzgerald-Flaujeac Factor; Fitzgerald Factor; HMWK-Kallikrein Factor. Accession Number: P08934. Expression Region: Leu214~Asp475. Tag Info: N-terminal His and GST Tag. Theoretical MW: 59kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.