Learn More
Biomatik Corporation Recombinant Indoleamine-2,3-Dioxygenase (IDO), 100UG
Catalog No. 501505586
Encompass_Preferred
Recombinant Human Indoleamine-2,3-Dioxygenase (IDO) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Indoleamine-2,3-Dioxygenase (IDO). Target Synonyms: CD107B; INDO; Indoleamine-Pyrrole 2,3 Dioxygenase. Accession Number: P14902. Gene ID: 3620. Expression Region: Ala2~Gly403. Tag Info: N-terminal His Tag. Theoretical MW: 43kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5586 Supplier Biomatik Corporation Supplier No. RPU41981
Recombinant Human Indoleamine-2,3-Dioxygenase (IDO) is a purified Prokaryotic Protein. Purity: >97% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Indoleamine-2,3-Dioxygenase (IDO). Target Synonyms: CD107B; INDO; Indoleamine-Pyrrole 2,3 Dioxygenase. Accession Number: P14902. Gene ID: 3620. Expression Region: Ala2~Gly403. Tag Info: N-terminal His Tag. Theoretical MW: 43kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.