Learn More
Biomatik Corporation Recombinant Inducible T-Cell Co Stimulator (ICOS), 100UG
Supplier:  Biomatik Corporation RPU41985
Encompass_Preferred
Recombinant Human Inducible T-Cell Co Stimulator (ICOS) is a purified Prokaryotic Protein. Purity: >90% by SDS-PAGE. Host: E. coli. Endotoxin Level: <1.0EU per 1ug, by the LAL method. Species: Human (Homo sapiens). Target Name: Inducible T-Cell Co Stimulator (ICOS). Target Synonyms: CD278; AILIM; Inducible T-Cell Costimulator; Activation-Inducible Lymphocyte Immunomediatory Molecule. Accession Number: Q9Y6W8. Gene ID: 29851. Expression Region: Phe31~Lys190. Tag Info: N-terminal His and GST Tag. Theoretical MW: 48kDa. Buffer: PBS, pH7.4, containing 0.01% SKL, 5% Trehalose. Storage: -20°C. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-5587